Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSPD™ Substrate (0.4 mM Ready-To-Use) with Emerald-II™ Enhancer, Size A

Catalog No. T2212S
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2212S 100 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2212S Supplier Invitrogen™ Supplier No. T2212
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready-to-use chemiluminescent alkaline phosphatase substrate, for protein or nucleic acid blotting on nitrocellulose membranes or solution-based assays.

Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP). Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.

  • Detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed and ease
  • Exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting
  • Use in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays
  • Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours
  • Enhancers increase light output and sensitivity
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Substrate
Quantity 100 ml
Detection Method Chemiluminescence
Form Liquid
Format Bottle
Product Line CSPD, NovaBright
Substrate Type Alkaline Phosphatase
Content And Storage Store the product at 4°C.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.