Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CST2 Monoclonal Antibody (OTI2D6), TrueMAB™, OriGene
SDP

Catalog No. 502540252
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0252 30 μL
50-167-5218 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0252 Supplier OriGene Supplier No. TA504560S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CST2 Monoclonal antibody specifically detects CST2 in Human, Mouse samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen CST2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2D6
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Ppat
Gene Accession No. Q06203
Gene Alias CST2; cystatin 2; cystatin S5; cystatin SA; cystatin-2; Cystatin-S5; cystatin-SA; cysteine-proteinase inhibitor; salivary cysteine (thiol) protease inhibitor
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 42-278 of human PPAT produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231327, 5471
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.