Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CstF-64 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987652
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7652 Supplier Bioss Supplier No. BS6995R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CSTF2 is one of three (including CSTF1 and CSTF3) cleavage stimulation factors which combine to form CSTF which is involved in the polyadenylation and 3′end cleavage of pre-mRNAs. CSTF2 contains a ribonucleoprotein-type RNA binding domain. CSTF2 is upregulated during activation of B cells which results in the switch of IgM heavy chain mRNA from membrane bound form to the secreted form.

Specifications

Antigen CstF-64
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CSTF2
Gene Accession No. P33240
Gene Alias alphaCstF-64 variant 4; betaCstF-64 variant 1; betaCstF-64 variant 2; betaCstF-64 variant 3; C630034J23Rik; CF-1 64 kDa subunit; Cleavage stimulation factor 64 kDa subunit; Cleavage stimulation factor subunit 2; cleavage stimulation factor, 3′ pre-RNA subunit 2; cleavage stimulation factor, 3′ pre-RNA subunit 2, 64 kDa; cleavage stimulation factor, 3′ pre-RNA, subunit 2; cleavage stimulation factor, 3′ pre-RNA, subunit 2, 64kD; cleavage stimulation factor, 3′ pre-RNA, subunit 2, 64kDa; CstF 64; CSTF 64 kDa subunit; Cstf2; Cstf64; CstF-64; mir-3112
Gene Symbols CSTF2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CSTF2T/CstF-64.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1478
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.