Learn More
Aviva Systems Biology Corp CSTF3 Antibody - N-terminal region (ARP40354_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40354_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSTF3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CSTF3. Alternate names: CSTF-77ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57407