Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific CT-Caps for CapTrack™

Catalog No. 22171739
Change view
Click to view available options
Color:
Black
Blue
Clear
Green
Lavender
Orange
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-171-739 Red
22-171-738 Orange
22-171-741 Yellow
22-171-735 Green
22-171-733 Blue
22-171-737 Lavender
22-171-734 Clear
22-171-740 White
22-171-736 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 22-171-739 Supplier Globe Scientific Supplier No. CTCAPR
Add to Cart
Add to Cart

Globe Scientific™ CT-Caps for CapTrack™ are available in a variety colors.

Specifications

Closure Size 13 mm
Autoclavable Not Autoclavable
Color Red
Quantity 1000 Caps
Sterility Non-sterile
For Use With (Equipment) CapTrack™
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.