Learn More
CTI Impervious Polylatex Shoe Covers
Description
- Help to control contamination
- Non-skid
- Impervious to liquids
- Individually inspected for stitching, sizing, materials and workmanship
Specifications
Specifications
| Size | Large |
| Material | Polylatex |
| Latex Free | No |
| Color | White |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Packaging Quantity | 150/Case (3 Packs of 50) |
| Product Type | Controlled Environments Shoe Covers |