Learn More
CTI Impervious Polylatex Shoe Covers
Catalog No. 19121103E
Change view
Click to view available options
Color:
Blue
White
Size:
2X-Large
3X-Large
Large
Universal
Packaging Quantity:
150/Case (3 Packs of 50)
300 Pairs/Cs.
300/Cs.
500 Pairs/Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size Packaging Quantity
19-121-103E White 2X-Large 300 Pairs/Cs.
19-099-014 White Universal 500 Pairs/Cs.
19-098-874 White Large 150/Case (3 Packs of 50)
19-121-103F Blue 3X-Large 300/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-121-103E Supplier CTI Supplier No. SCPL150HD
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Strong and durable with a comfortable fit. Compatible with ISO 6 (Class 1,000) environments.
  • Help to control contamination
  • Non-skid
  • Impervious to liquids
  • Individually inspected for stitching, sizing, materials and workmanship
Specifications
Size 2X-Large
Material Polylatex
Latex Free No
Color White
Disposable Single-use
Sterility Non-sterile
Thickness 4.7 mil
Packaging Quantity 300 Pairs/Cs.
Dimensions (L x W) 16 x 6 in. (40.64 x 15.24cm)
Length (English) 16 in.
Length (Metric) 40.64 cm
Product Type Controlled Environments Shoe Covers