Learn More
Specifications
Specifications
| For Use With (Application) | Antibody Competition |
| Formulation | PBS and 1 M Urea, pH 7.4. |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 26 kDa |
| Quantity | 0.1 mL |
| Storage Requirements | Store at -20°C. Avoid freeze/thaw cycles. |
| Gene Alias | 114 kDa Rho-specific guanine nucleotide exchange factor, rho guanine nucleotide exchange factor 18, Rho/Rac guanine nucleotide exchange factor (GEF) 18, Rho-specific guanine nucleotide exchange factor p114, RP78; SA-RhoGEF; p114RhoGEF; P114-RhoGEF, septin-associated RhoGEF |
| Inhibitor Properties | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-48929. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mLFor current lot information, including availability, please contact our technical support team click nb-technical@bio-techne.com For further blocking peptide related information and a protocol, click here . |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Protein | CTB-133G6.1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
