Learn More
Invitrogen™ CTC1 Polyclonal Antibody

Description
CTC1 Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | CTC1 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CTC1 |
| Gene Accession No. | Q2NKJ3 |
| Gene Alias | AAF132; AAF-132; alpha accessory factor 132; C17orf68; conserved telomere capping protein 1; Conserved telomere maintenance component 1; CRMCC; CST complex subunit CTC1; CTC1; CTS telomere maintenance complex component 1; HBV DNAPTP1-transactivated protein B; tmp494178 |
| Gene Symbols | CTC1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.