Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD CTGCTV2 for BD COR™ System

Catalog No. 21564101 Shop All Waters Corporation Products
Change view
Click to view available options
Type:
Assay Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
21-564-101 Assay Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-564-101 Supplier BD Supplier No. 445435
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

FDA - Cleared assays that combines the 3 most prevalent non-viral STIs in one test for asymptomatic and symptomatic individuals

  • 3-in-1 assay for the 3 most common non-viral STIs
    • Chlamydia trachomatis (CT)
    • Neisseria gonorrhoeae (GC)
    • Trichomonas vaginalis (TV)
  • Approved for Symptomatic and Assymptomatic testing
  • Sample types include:
    • Urine - CT, GC, TV
    • Liquid-Based Cytology (LBC) specimens - CT, GC
    • Endocervical Swab - CT, GC
    • Vaginal Swab - Self Collected - CT, GC, TV
    • Vaginal Swab - Clinician Collected - CT, GC, TV
    • Rectal Swab - Clinician Collected - CT, GC
    • Oropharyngeal Swab - Clinician Collected - CT, GC
  • Includes Trichomonas vaginalis infections that tend to be asymptomatic
  • Includes dual gene targets for GC, which maximizes the assay specificity: both GC gene targets are required for a positive GC result
  • Compatible with the BD Vaginal Panel for BD COR™ System for comprehensive testing of vaginitis and STIs
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 27°C
Type Assay Kit
Includes Master Mix, Liquid Extraction Reagent Plate
Quantity 1 Pk.
Dimensions (L x W x H) 32 x 29 x 33 mm
Weight (English) 5 lb.
Electrical Requirements None
Form Ready-to-Use (RTU)
Operating Temperature Range Up to 32°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.