Learn More
FUJIFILM IRVINE SCIENTIFIC INC CTGrade GMP rh FLT3 Lgnd 100UG
Supplier: FUJIFILM IRVINE SCIENTIFIC INC 50003100UG
FLT-3 Ligand is a growth factor that regulates proliferation of early hematopoietic cells. FLT-3 Ligand binds to cells expressing the tyrosine kinase receptor FLT-3. FLT-3 Ligand by itself does not stimulate proliferation of early hematopoietic cells, but synergizes with other CSFs and interleukins to induce growth and differentiation Unlike SCF, FLT-3 Ligand exerts no activity on mast cells. Multiple isoforms of FLT-3 Ligand have been identified The predominant biologically active form is anchored to the cell surface as the extracellular domain of a transmembrane protein (209 aa) The membrane-bound isoform can be proteolytically cleaved to generate a biologically active soluble isoformThe biological activity of CTGrade GMP interleukins and growth factors is standardized, where applicable, to WHO International standards, providing cell and gene therapy developers consistent, lot-to-lot biological activity and performance.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.