Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTI Industrial Powder Free Nitrile Gloves

Catalog No. 19050354
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-050-354 2X-Large
19-050-350 Small
19-050-351 Medium
19-050-352 Large
19-050-353 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-050-354 Supplier CTI Supplier No. NPXINBLK105
This item is not returnable. View return policy

NPXINBLK series nitrile gloves are black in color and feature textured finger tips and palms for use in industrial applications.

  • Black, soft feel
  • Textured finger tips and palm for secure grip
  • Tapered, beaded cuff
  • Latex-free
  • 10″ Length, Ambidextrous
  • Leak tested

Specifications

Certifications/Compliance FDA 21 CFR 177.2600, ASTM D3578-05, MIL STD 105E
Applications Industrial
AQL 1.5
Color Black
Exterior Finish Fully Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size 2X-Large
Thickness (English) 5 mil
Thickness (Metric) 0.12 mm
Model NPXINBLK105
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.