Learn More
                                    CPC Scientific H-DPhe-Cys-Tyr-DTrp-Orn-Thr-Pen-Thr-ol (trifluoroacetate salt) 1MG
Supplier: CPC Scientific SOMA006B
            Somatostatin analog CTOP is a very potent and selective ligand for µ-opioid receptors.ONE-LETTER SEQUENCE: fCYwOT-Pen-T-olMOLECULAR FORMULA: C50H67N11O11S2MOLECULAR WEIGHT:1062.28STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [103429-31-8], SYNONYMS: (Tyr3,Orn5,Pen7)-Octreotate amide, NTB (Naltriben)RESEARCH AREA: Pain
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.