Learn More
Abcam CTPB, p300 HAT Activator, Benzamide, 554.1 Da, 5 mg
Description
- Compound class: Benzamide
- Activity: Small-molecule p300 histone acetyltransferase (HAT) activator; active in vitro and in vivo
- Purity: >99%
- Molecular weight: 554.1 Da
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | p300 Histone Acetyltransferase |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Form | Solid |
| Buffer | 100% HAT activator, CTPB |
| Content And Storage | Store at -20°C. |
| Format | Vial |
| Formulation | 100% HAT Activator, CTPB |
| Quantity | 5 mg |
| For Use With (Application) | p300 histone acetyltransferase activation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.