Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. APPA6586
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APPA6586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APPA6586 Supplier Invitrogen™ Supplier No. PA116586
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: human breast cancer and kidney tissue.

The trace metal copper (Cu) plays a crucial role in mammalian cells as a cofactor for many enzymes. Cu-related genetic diseases, such as Menkes disease and Wilson disease, arise from a lack of Cu homeostasis in mammalian cells. CTR1 is a high-affinity copper-uptake protein. The C-terminal domain is similar to the Raf family of protein kinases, but it's first two-thirds encodes a novel protein domain. CTR1 provides the primary avenue for copper uptake in mammalian cells, thereby, affecting Cu homeostasis and embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTR1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SLC31A1
Gene Accession No. O15431, Q8K211, Q8WNR0, Q9JK41
Gene Alias 4930445G01Rik; AI787263; AU016967; copper transport 1 homolog; copper transporter 1; Copper uptake transporter 1; COPT1; ctr1; ctr-1; hCTR1; high affinity copper uptake protein; high affinity copper uptake protein 1; high-affinity copper uptake protein; liver regeneration-related protein LRRGT00200; LRRGT00200; rCTR1; SLC31A1; slc31a1.L; solute carrier family 13 (sodium/sulphate symporters), member 1; solute carrier family 31 (copper transporter), member 1; solute carrier family 31 (copper transporter), member 1 L homeolog; solute carrier family 31 (copper transporters), member 1; solute carrier family 31 member 1; solute carrier family 31, member 1; Xctr1; XELAEV_18038194mg; zgc:76839
Gene Symbols SLC31A1, slc31a1.L
Host Species Rabbit
Immunogen A synthetic peptide derived from a C-terminal sequence of human SLC31A1/CTR1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1317, 171135, 20529, 379422, 397186, 403028, 481678
Target Species Human, Mouse, Rat, Canine, Pig, Frog, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.