Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTR1 Recombinant Rabbit Monoclonal Antibody (23GB4635), Invitrogen™
GREENER_CHOICE

Catalog No. MA552720
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552720 100 μL
MA552721 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552720 Supplier Invitrogen™ Supplier No. MA552720
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

The trace metal copper (Cu) plays a crucial role in mammalian cells as a cofactor for many enzymes. Cu-related genetic diseases, such as Menkes disease and Wilson disease, arise from a lack of Cu homeostasis in mammalian cells. CTR1 is a high-affinity copper-uptake protein. The C-terminal domain is similar to the Raf family of protein kinases, but it's first two-thirds encodes a novel protein domain. CTR1 provides the primary avenue for copper uptake in mammalian cells, thereby, affecting Cu homeostasis and embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTR1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4635
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC31A1
Gene Accession No. O15431, Q8K211, Q9JK41
Gene Alias 4930445G01Rik; AI787263; AU016967; copper transport 1 homolog; copper transporter 1; Copper uptake transporter 1; COPT1; ctr1; ctr-1; hCTR1; high affinity copper uptake protein; high affinity copper uptake protein 1; high-affinity copper uptake protein; liver regeneration-related protein LRRGT00200; LRRGT00200; rCTR1; SLC31A1; slc31a1.L; solute carrier family 13 (sodium/sulphate symporters), member 1; solute carrier family 31 (copper transporter), member 1; solute carrier family 31 (copper transporter), member 1 L homeolog; solute carrier family 31 (copper transporters), member 1; solute carrier family 31 member 1; solute carrier family 31, member 1; Xctr1; XELAEV_18038194mg; zgc:76839
Gene Symbols SLC31A1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CTR1/SLC31A1 (1-32AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1317, 171135, 20529
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.