Learn More
New England Biolabs, Inc. Control LAMP Primer Mix (rActin) - 50 rxns
SDP
Catalog No. 502237903
Encompass
The Control LAMP Primer Mix (rActin) targets actin RNA at an exon-exon junction. It is a LAMP primer set (F3 B3 FIP BIP LF LB) that amplifies rActin to ensure the absence of inhibition from human nucleic acid templates in a LAMP reaction. Control LAMP Primer Sequences rActin (5 3) rActin Primer Set Sequence ACTB-F3 AGTACCCCATCGAGCACG ACTB-B3 AGCCTGGATAGCAACGTACA ACTB-FIP GAGCCACACGCAGCTCATTGTATCACCAACTGGGACGACA ACTB-BIP CTGAACCCCAAGGCCAACCGGCTGGGGTGTTGAAGGTC ACTB-LF TGTGGTGCCAGATTTTCTCCA ACTB-LB CGAGAAGATGACCCAGATCATGT
This item is not returnable. View return policy
Catalog No. 50-223-7903 Supplier New England Biolabs, Inc. Supplier No. S0164S
The Control LAMP Primer Mix (rActin) targets actin RNA at an exon-exon junction. It is a LAMP primer set (F3 B3 FIP BIP LF LB) that amplifies rActin to ensure the absence of inhibition from human nucleic acid templates in a LAMP reaction. Control LAMP Primer Sequences rActin (5 3) rActin Primer Set Sequence ACTB-F3 AGTACCCCATCGAGCACG ACTB-B3 AGCCTGGATAGCAACGTACA ACTB-FIP GAGCCACACGCAGCTCATTGTATCACCAACTGGGACGACA ACTB-BIP CTGAACCCCAAGGCCAACCGGCTGGGGTGTTGAAGGTC ACTB-LF TGTGGTGCCAGATTTTCTCCA ACTB-LB CGAGAAGATGACCCAGATCATGT