Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CTS™ Viral-Plex™ Complexation Buffer

Catalog No. A5145402
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 L
10 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5145402 1 L
A5145401 500 mL
A5145403 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A5145402 Supplier Gibco™ Supplier No. A5145402
Add to Cart
Add to Cart

Gibco CTS Viral-Plex Complexation Buffer is a chemically defined, protein-free, phenol-red-free buffer for complexing plasmid DNA with a transfection reagent for standard transfection applications.

Gibco CTS Viral-Plex Complexation Buffer is a chemically defined, protein-free, phenol-red-free buffer for complexing plasmid DNA with a transfection reagent for standard transfection applications. It is a part of the CTS AAV-MAX system and is designed to be used with the CTS AAV-MAX Transfection Kit for the transient transfection of CTS Viral Production Cells 2.0.

This buffer is manufactured in conformity with cGMP for medical devices (21 CFR Part 820) and follows USP <1043> and European Pharmacopoeia (Ph. Eur.) 5.2.12 recommendations. A Drug Master File and Regulatory Support File are available upon request.

The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade (Cat. No. A51217) and GMP versions to enable seamless transition from research to commercial manufacturing. Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0 (A48400), CTS Viral Production Medium (A5144001), CTS AAV-MAX Transfection Kit (A5427701), and Viral Production Medium, AGT (A5147101).

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Transient transfection for mammalian AAV production
Physical Form Liquid
Quantity 1 L
Packaging Bottle
Sterility Sterile-filtered
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.