Learn More
Bioworld Cu-Alginate Macrobeads (BAB), 100 mL
Supplier:  Bioworld 201100032
Encompass_Preferred
Bioalginate beads Alginate, commonly called sodium alginate, is a linear polysaccharide. The copolymer consists of two uronic acids: D-mannuronic acid (M) and L-guluronic acid (G ). Sodium alginate as well as other alkali metal alginates are water soluble, whereas the salts of polyvalent cations, e.g., calcium, nickel, copper, cobalt are water insoluble, with the exception of magnesium. Polyvalent cations bind to the polymer whenever there are two neighboring guluronic acid residues. Thus, polyvalent cations are responsible for the cross-linking of both different polymer molecules and different parts of the same polymer chain. The process of gelation, i.e. the exchange of polyvalent ions for sodium ions is carried out under relatively mild conditions. Technical specificationsMicrobeads: 150-250 µm. Used for protein fractionation & affinity chromatography. Macrobeads: 500µm-3mm. Used for enzymes and cell mobilization
Catalog No. 50-197-501
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.