Learn More
TARGETMOL CHEMICALS INC CU-CPD107 5MG
Supplier: TARGETMOL CHEMICALS INC T35349
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. CU-CPD107 is a selective agonist of toll-like receptor 8 (TLR8) and also shows activation activity against ssRNA ligands. CU-CPD107 acts as an inhibitor of TLR8 signaling in the presence of R848 (IC50=13.7 uM). CU-CPD107 exhibited coagonist activity in the presence of ssRNA while CU-CPD107 alone could not affect TLR8 signal transduction. purity: 98%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.