Learn More
TARGETMOL CHEMICALS INC BAY-3827 5MG
Supplier: TARGETMOL CHEMICALS INC T733505MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. BAY-3827 is a potent and selective AMPK inhibitor with potential antitumor activity, demonstrating antiproliferative effects in an androgen-dependent prostate cancer model and inhibiting the phosphorylation of acetyl CoA carboxylase 1 (ACC-1). Purity 100%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.