Learn More
TARGETMOL CHEMICALS INC CU-CPT-9A 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T7317
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CU-CPT-9a is a potent TLR8 inhibitor (IC50 0.5 nM) that suppresses TLR8-mediated proinflammatory signaling in various cell lines and human primary cells-purity: 98%
Catalog No. 50-257-6329
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.