Learn More
Selleck Chemical LLC 4-Methyl-2-oxovaleric acid S2987-1g
SDP
Supplier:  Selleck Chemical LLC S29871G
Encompass_Preferred
4-Methyl-2-oxovaleric acid (Ketoleucine 4-MOV KIC 4-Methyl-2-oxopentanoic acid alpha-Ketoisocaproic acid alpha-ketoisocaproate 2-Oxoisohexanoate) is released by Astrocytes to neurons and can be reaminated by aminotransferase to leucine 4-Methyl-2-oxovaleric acid reduces the rate of protein degradation in skeletal muscle 4-Methyl-2-oxovaleric acid acts as an uncoupler of oxidative phosphorylation (OXPHOS) and as a metabolic inhibitor possibly through its inhibitory effect on alpha-ketoglutarate dehydrogenase (oxoglutarate dehydrogenase complex OGDC) activity
Catalog No. 50-311-2431
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.