Learn More
Sartorius Cubis™ II 36P High-Capacity Essential Micro Balance
Catalog No. 01675447 Shop All Sartorius Products
Change view
Click to view available options
Model:
Standard Configuration with Draft Shield
Catalog No. Model
01-675-447 Standard Configuration with Draft Shield
1 options
Catalog No. 01-675-447 Supplier Sartorius Supplier No. MCE36P3S00D
The Cubis™ II range of high-capacity micro balances with a maximum load between 32 g and 111 g and a readability between 0.001 mg and 0.002 mg provide the ideal choice for a broad range of applications.
  • Ultra-high resolution with up to 61 million weighing steps (digits)
  • End-to-end data integrity. 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management
  • Cleanability, part of compliance
  • Error-free operation. Individual QApp workflows, motorized auto-leveling
  • Allow upgrades for hardware features like automated draft shields or built-in ionizer
  • Optional inner draft shield for best weighing performance
  • Housing: Stainless steel 1.4401, 1.4404, Aluminum, Plastic PBT, PA, Float glass Optiwhite, Control Unit: Aluminum, painted, Plastic PBT, PP, Float glass
TRUSTED_SUSTAINABILITY
Specifications
Connection Type USB A, USB B, USB C
Capacity 32 g, 10.1 g
Display Type Color TFT Touchscreen
Readability 0.01 mg, 0.001 mg
Material Stainless Steel
Model Standard Configuration with Draft Shield
Stabilization Time 3.5 sec., 2.5 sec.
Type Micro Balance
Electrical Requirements 100/240 V
Legal for Trade No