Learn More
Sartorius Cubis™ II 36S High-Capacity Essential Micro Balance
GREENER_CHOICE
Catalog No. 01675443 Shop All Sartorius Products
Change view
Click to view available options
Model:
HWL Configuration with Draft Shield
ION Configuration with Draft Shield
MDS Configuration with Draft Shield
Standard Configuration with Draft Shield
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
01-675-443 Standard Configuration with Draft Shield
01-675-440 MDS Configuration with Draft Shield
01-675-441 ION Configuration with Draft Shield
01-675-442 HWL Configuration with Draft Shield
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-675-443 Supplier Sartorius Supplier No. MCE36S3S00D
Only null left
Add to Cart
Add to Cart
Request a Quote
The Cubis™ II range of high-capacity micro balances with a maximum load between 32 g and 111 g and a readability between 0.001 mg and 0.002 mg provide the ideal choice for a broad range of applications.
  • Ultra-high resolution with up to 61 million weighing steps (digits)
  • End-to-end data integrity. 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management
  • Cleanability, part of compliance
  • Error-free operation. Individual QApp workflows, motorized auto-leveling
  • Allow upgrades for hardware features like automated draft shields or built-in ionizer
  • Optional inner draft shield for best weighing performance
  • Housing: Stainless steel 1.4401, 1.4404, Aluminum, Plastic PBT, PA, Float glass Optiwhite, Control Unit: Aluminum, painted, Plastic PBT, PP, Float glass
TRUSTED_SUSTAINABILITY
Specifications
Connection Type USB A, USB B, USB C
Capacity 32 g
Display Type Color TFT Touchscreen
Readability 0.001 mg
Material Stainless Steel
Model Standard Configuration with Draft Shield
Stabilization Time 3.5 sec.
Type Micro Balance
Electrical Requirements 100/240 V
Legal for Trade No