Learn More
Sartorius Cubis™ II 66P High-Capacity Advanced Micro Balance
GREENER_CHOICE
Catalog No. 01675463 Shop All Sartorius Products
Change view
Click to view available options
Model:
HWL Configuration with Draft Shield
ION Configuration with Draft Shield
MDS Configuration with Draft Shield
QP99 Configuration with Draft Shield
QP99 HWL Configuration with Draft Shield
QP99 ION Configuration with Draft Shield
QP99 MDS Configuration with Draft Shield
Standard Configuration with Draft Shield
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
01-675-463 Standard Configuration with Draft Shield
01-675-460 MDS Configuration with Draft Shield
01-675-461 ION Configuration with Draft Shield
01-675-462 HWL Configuration with Draft Shield
01-675-459 QP99 Configuration with Draft Shield
01-675-456 QP99 MDS Configuration with Draft Shield
01-675-457 QP99 ION Configuration with Draft Shield
01-675-458 QP99 HWL Configuration with Draft Shield
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 01-675-463 Supplier Sartorius Supplier No. MCA66P3S00D
Only null left
Add to Cart
Add to Cart
Request a Quote
The Cubis™ II range of high-capacity micro balances with a maximum load between 32 g and 111 g and a readability between 0.001 mg and 0.002 mg provide the ideal choice for a broad range of applications.
  • Ultra-high resolution with up to 61 million weighing steps (digits)
  • End-to-end data integrity. 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management
  • Cleanability, part of compliance
  • Error-free operation. Individual QApp workflows, motorized auto-leveling
  • Allow upgrades for hardware features like automated draft shields or built-in ionizer
  • Optional inner draft shield for best weighing performance
  • Housing: Stainless steel 1.4401, 1.4404, Aluminum, Plastic PBT, PA, Float glass Optiwhite, Control Unit: Aluminum, painted, Plastic PBT, PP, Float glass
TRUSTED_SUSTAINABILITY
Specifications
Connection Type USB A, USB B, USB C
Capacity 61 g, 12 g
Display Type Color TFT Touchscreen
Readability 0.01 mg, 0.001 mg
Material Stainless Steel
Model Standard Configuration with Draft Shield
Stabilization Time 3.5 sec., 2.5 sec.
Type Micro Balance
Electrical Requirements 100/240 V
Legal for Trade No