Learn More
Sartorius Cubis™ II Balance PROMO
Catalog No. 01672090PM Shop All Sartorius Products
Change view
Click to view available options
Capacity:
220 g
Catalog No. Capacity
01-672-090PM 220 g
1 options
Catalog No. 01-672-090PM Supplier Sartorius Supplier No. MCA225SI3MTSPM
The Cubis™ II can be individually configured from different components. The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a range of accessories allows the individual adaptation to all weighing tasks.
  • Leading performance. Monolithical weighing system, integrated climate sensors, individual sample holders Error-free operation. Individual QApp workflows, motorized auto-leveling
  • Full end-to-end data integrity. 21 CFR Part 11 compli-ance, integrated audit-trail, state-of-the-art user management
  • Outstanding service support. Integrated status center, integrated service functions preventive maintenance on accredited standards
TRUSTED_SUSTAINABILITY
Specifications
Capacity 220 g
Display Type Color Touchscreen
Readability 0.01 mg
Model MCA with Draft Shield D
Type Balance