Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Cubis II™ Essential High Capacity Balance
GSA_VAGREENER_CHOICE

Catalog No. 14561208 Shop All Sartorius Products
Change view
Click to view available options
Model:
36200S
36201P
Readability:
0.1 g, 1 g
1 g
Capacity:
36.2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Readability Capacity
14-561-208 36201P 0.1 g, 1 g 36.2 kg
14-561-212 36200S 1 g 36.2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-561-208 Supplier Sartorius Supplier No. MCE36201P2S000
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

Cubis II™ Essential High Capacity Balance offers a high level of weighing accuracy with a large, high-contrast touch display that features factory-installed essential weighing applications for easy operation.

  • Top Features of the Cubis II™ Essential High-Capacity Balance
  • Stainless steel weigh pan
  • Convenient essential user interface
  • 1 g readability
  • Popular Applications for the Cubis II™ Essential High-Capacity Balance: Large sample weighing, Chemical analysis and consumer care

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Connection Type USB A USB B USB C RS-232 9 Pin peripheral connector for Sartorius accessories
Capacity 36.2 kg
Display Type Touchscreen
Readability 0.1 g, 1 g
Material Lead (CAS 7439-92-1): This substance is present as an ingredient in brass alloys. It complies with the limit of 0.4% by weight of EU Directive 2011/65 / EU (RoHS) in these alloys. Bisphenol A (CAS 80-05-7): Bisphenol A is the raw material for the plastic polymers polycarbonate and polysulfone. Sartorius uses these two polymers in some plastic components. Bisphenol A is firmly bound in the structure of the plastic. If used as intented, no adverse health effects are to be expected.
Model 36201P
Stabilization Time 1.5 sec.
Type High Capacity Toploader
Electrical Requirements CR2030
Legal for Trade Class III
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.