Learn More
Market Lab Cubitainer Carts
Supplier: Market Lab 2439
The Cubitainer Cart from MarketLab is a utility cart designed to transport and dispense liquids from cubitainers. It features a sturdy steel frame with 4" casters for easy mobility. The top shelf has cutouts to hold cubitainers securely in place while dispensing liquid through the spigot, and the lower shelf provides storage space. This utility cart allows convenient transport and dispensing of liquids in industrial, commercial, and institutional settings. With a 150 lb weight capacity per shelf, this cart is a durable and functional solution for liquid handling needs.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.