Learn More
Market Lab Cubitainer Organizer Stand, ML2377
Supplier:  Market Lab 2377
Encompass_Preferred
The Cubitainer Organizer Stand by MarketLab is a metal stand designed to hold and dispense cubitainer bags. It features sturdy steel construction with powder coated finish and can hold up to 4 cubitainer bags at a time. The stand has 4 removable hooks to hang bags and a drip tray to catch leaks and spills. It measures 23"W x 19.5"D x 51.5"H and has non-skid rubber feet for stability. The Cubitainer Organizer Stand provides an organized dispensing solution for cubitainer bags.
Catalog No. NC9914156
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.