Learn More
TARGETMOL CHEMICALS INC CUCURBIT8URIL 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T35862
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Cucurbit[8]uril is a highly effective and safe supramolecular compound that promotes protein heterodimerization. It selectively induces the heterodimerization of methylviologen and naphthalene functionalized proteins demonstrating its specificity and versatility. Additionally Cucurbit[8]uril shows exceptional oral activity and low toxicity making it a promising candidate for various pharmaceutical and biotechnological applications. purity: 98%
Catalog No. 50-258-0207
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.