Learn More
Cayman Chemical TetrahydroepIberberIn 25mg
SDP
Supplier:  Cayman Chemical 3617725 MG
Encompass_Preferred
An IGF-1R inhibitor (IC50 0 15 UM in a cell-free assay) selective for IGF-1R over a panel of 20 kinases but also inhibits InsR FLT1 FLT3 and Tie2 (IC50s 0 14 0 6 0 42 and 0 53 UM respectively in cell-free assays) selectively inhibits the autophosphorylation of IGF-1R over InsR EGFR PDGFR c-Kit and Bcr-Abl in cells (IC50s 0 086 2 3 10 10 5 and 10 UM respectively) reduces tumor volume in an NWT-21 murine fibrosarcoma model at 20 30 or 50 mg/kg twice per day
Catalog No. 50-291-1683
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.