Learn More
Medchemexpress LLC 7-(4-(3-ethynylphenylamino)-7-methoxyquinazolin-6-yloxy)-N-hydroxyheptanamide | 1012054-59-9 | MFCD15528940 | 99.1% | 434.49 g/mol | C24H26N4O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1022310MM 1ML
Encompass_Preferred
CUDC-101 is a small-molecule inhibitor that targets histone deacetylases (HDAC) and receptor tyrosine kinases EGFR and HER2. It is intended for research use in studies of signaling pathways, anti-proliferative activity, and preclinical cancer models. The compound is supplied as a high-purity reagent suitable for in vitro experiments.
  • Multi-target HDAC, EGFR, and HER2 inhibition
  • High purity suitable for research applications
  • Available as a ready-to-use 10 mM solution in DMSO (1 mL)
  • Confirmed molecular formula and molecular weight for analytical workflows
  • Intended for in vitro and preclinical research use
Catalog No. 50-002-47089