Learn More
Apexbio Technology LLC Belinostat (PXD101) 414864-00-9 200mg
Supplier: Apexbio Technology LLC A4096200
Belinostat (PXD101) is a hydroxamic acid-based inhibitor targeting histone deacetylase (HDAC) enzymes Its mechanism involves the suppression of HDAC enzymatic activity leading to increased acetylation of histone proteins H3 and H4 within cells In biochemical assays using Hela cell extracts belinostat exhibits inhibitory activity against HDAC with an IC50 value of approximately 27 nM In tumor cell culture models belinostat induces cytotoxicity and represses cellular proliferation with reported IC50 values ranging roughly from 0 5 M to 10 M varying by cell type and origin making it suitable for research on epigenetic modulation in oncology specifically in studies of bladder prostate and related cancer cell lines
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.