Learn More
Medchemexpress LLC 7-((4-(3-ethynylphenylamino)-7-methoxyquinazolin-6-yl)oxy)-N-hydroxyheptanamide | 1012054-59-9 | MFCD15528940 | >98.0% | 434.49 g/mol | C24H26N4O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1022350MG
Encompass_Preferred
CUDC-101 is a small-molecule research inhibitor of histone deacetylases (HDACs), epidermal growth factor receptor (EGFR), and HER2, used in preclinical cancer research to assess antiproliferative and target-specific effects.
  • Potent inhibitor of HDAC, EGFR, and HER2 with low-nanomolar activity.
  • Demonstrates broad antiproliferative effects across cancer cell lines.
  • Supplied as a light yellow to yellow powder for laboratory use.
  • High purity suitable for research applications (reported ≥98% by HPLC).
  • Available in small-mass packaging for research workflows.
  • Molecular weight 434.49 g/mol.
Catalog No. 50-002-46578
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.