Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CUGBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596484
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596484 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596484 Supplier Invitrogen™ Supplier No. PA596484
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RKLFIGMISK KCTENDIRVM FSSFGQIEEC RILRGPDGLS RGCAFVTFTT RAMAQTAIKA MHQAQTMEGC SSPMVVKFAD TQKDKEQKRM AQQLQQQMQQ ISAASVWGNL AGLNTLGPQY LALLQQTASS GNLNTLSSLH PMGGLNAMQL QNLAALAAAA SAAQNTPSGT NALTTSSSPL SVLTSSAGSS PSSSSSNSVN PIASLGALQT LAGATAGLNV GSLAGMAALN GGLGSSGLSN GTGSTMEALT QAYSGIQQYA AAALPTLYNQ NLLTQQSIGA AGSQKEGPEG ANLFIYHLPQ EFGDQDLLQM FMPFGNVVSA KVFIDKQTNL SKCFGFVSYD NPVSAQAAIQ SMNGFQIGMK RLKVQLKRSK NDSKPY; Positive Samples: U-87MG, SH-SY5Y, HepG2, NIH/3T3, HeLa, Mouse liver, Mouse testis; Cellular Location: Cytoplasm, Nucleus.

Myotonic dystrophy (MD) is an autosomal dominant neuromuscular disease that is associated with a (CTG)n repeat expansion in the 3'-untranslated region of the myotonin protein kinase (Mt-PK) gene. A (CUG) n oligonucleotides triplet repeat pre-mRNA/mRNA binding protein may play an important role in DM pathogenesis. HeLa cell protein, CUG-BP1, has been purified based upon its ability to bind specifically to (CUG) 8 oligonucleotides in vitro. CUG-BP1 is the major (CUG) 8 binding activity in normal cells. CUG-BP1 has been identified as isoforms of a novel heterogeneous nuclear ribonucleoprotein (hnRNP), hNab50. The CUG-BP/hNab50 protein is localized predominantly in the nucleus and is associated with polyadenylated RNAs in vivo. In vitro RNA-binding/photocrosslinking studies demonstrate that CUG-BP/hNab50 binds to RNAs containing the Mt-PK 3-UTR. The (CUG) n repeat region in Mt-PK mRNA is a binding site for CUG-BP/hNab50 in vivo, and triplet repeat expansion leads to sequestration of this hnRNP on mutant Mt-PK transcripts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CUGBP1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.70 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CELF1
Gene Accession No. P28659, Q4QQT3, Q92879
Gene Alias 1600010O03Rik; 50 kDa nuclear polyadenylated RNA-binding protein; AA407467; Brain protein F41; Brunol2; bruno-like 2; bruno-like protein 2; Celf1; CELF-1; CUG RNA-binding protein; CUG triplet repeat RNA-binding protein 1; CUG triplet repeat, RNA binding protein 1; CUG triplet repeat, RNA-binding protein 1; Cugbp; CUG-BP; CUG-BP- and ETR-3-like factor 1; CUGBP Ela; CUGBP Elav-like family member 1; CUGBP, Elav-like family member 1; CUGBP, Elav-like family member 1-like; CUGBP1; CUG-BP1; D2Wsu101e; Deadenylation factor CUG-BP; deadenylation factor EDEN-BP; EDEN-BP; EDEN-BP homolog; embryo deadenylation element binding protein; embryo deadenylation element-binding protein homolog; HNAB50; NAB50; NAPOR; nuclear polyadenylated RNA-binding protein, 50-kD; RNA-binding protein BRUNOL-2; RP23-147D3.7
Gene Symbols CELF1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 108-483 of human CELF1 (NP_9419891).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10658, 13046, 362160
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.