Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CUL4A Mouse anti-Human, Mouse, Non-human primate, Porcine, Rat, Clone: 1A7F12, Proteintech
SDP

Catalog No. p-7213442 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6478 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6478 Supplier Proteintech Group Inc Supplier No. 660381IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.

Specifications

Antigen CUL4A
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1A7F12
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CUL4A
Gene Accession No. Q13619, Q3TCH7
Gene Alias CUL 4A, CUL4A, cullin 4A
Gene Symbols Cul4a
Host Species Mouse
Immunogen CUL4A Fusion Protein Ag18089
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100517912, 361181, 8451, 99375
Target Species Human, Mouse, Non-human Primate, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.