Learn More
Invitrogen™ Cullin 4B Monoclonal Antibody (OTI1C4)

Description
Cullin 4B Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | Cullin 4B |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1C4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CUL4B |
| Gene Accession No. | A2A432, Q13620 |
| Gene Alias | 2700050M05Rik; AA409770; CUL4B; CUL-4B; cullin 4B; cullin-4B; EGK_20888; KIAA0695; mKIAA0695; MRXHF2; MRXS15; MRXSC; RGD1564494; SFM2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.