Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Cultrex 3D Culture Matrix Laminin I
GSA_VASDP

Catalog No. 344600501 Shop All Novus Products
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
344600501 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 344600501 Supplier R&D Systems™ Supplier No. 344600501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications: Bioactivity

  • Promotes cell attachment and spreading of MG63 human osteosarcoma cells
  • Used as a gel or media additive with other basement membrane components
  • Helps to study cellular growth and differentiation in three dimensions in vitro

Storage Buffer:

  • Dulbecco's Modified Eagle's medium containing 10μg/ml gentamycin sulfate and no phenol red

Sterility Testing:

  • No bacterial or fungal growth detected after incubation at 37°C for 14 days
  • No mycoplasma contamination detected by PCR

Order Info

The product is shipped with dry ice or equivalent. Upon receipt, store it immediately at the temperature recommended below

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Concentration 6 mg/mL
Product Type Laminin I extracellular matrix protein
For Use With (Application) Bioactivity
Product Line R&D Systems
Quantity 30 mg
Endotoxin Level <20.0EU per 1μg of the protein by the LAL method
Purity or Quality Grade ≥90% (SDS-PAGE)
Source Murine Engelbreth-Holm-Swarm (EHS) tumor.
Content And Storage Product is stable for a minimum of 3 months from date of shipment when stored at -80° C in a manual defrost freezer. For optimal stability, store at -80° C in aliquots. Avoid repeated freeze/thaw cycles.

For research use only. Not for use in diagnostic procedures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.