Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Cultrex 3D Culture Matrix Rat Collagen I
GSA_VASDP

Catalog No. 344702001 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
344702001 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 344702001 Supplier R&D Systems™ Supplier No. 344702001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications: Bioactivity

Functional Assays: Promotes cell attachment and spreading of HT-1080 human fibrosarcoma cells. Collagen I promotes attachment and growth of murine endothelial SVEC4-10 cells in 3-D culture. Gelling Assay: Type I collagen forms a firm gel at neutral pH and 37°C when diluted to 0.4 mg/ml.

Order Info

The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Collagen I insoluble extracellular glycoproteins
Product Line R&D Systems
Quantity 100 mg
Endotoxin Level <20.0EU per 1μg of the protein by the LAL method
Regulatory Status RUO
Source Rat tail tendons
Content And Storage Product is stable for a minimum of 3 months if stored at 4 °C. Do Not Freeze. Storage Buffer: 20 mM Acetic Acid Sterility Testing: No bacterial or fungal growth detected after incubation at 37 °C for 14 days following USP XXIV Chapter 71 sterility test. No mycoplasma contamination detected by PCR.

For Research Use Only. Not For Use In Diagnostic Procedures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.