Learn More
Description
- Structural component of extracellular matrices found in connective tissue and internal organs
- Composed of two alpha1(I) chains and one alpha2(I) chain which forms a triple helix scaffold at a neutral pH and 37°C
Sterility Testing:
- No bacterial or fungal growth detected after incubation at 37°C for 14 days
- No mycoplasma contamination detected by PCR
Storage Buffer:
- 20mM Acetic Acid
Order Info
The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below
Specifications
Specifications
| Content And Storage | Product is stable for a minimum of 3 months if stored at 47°C |
| Product Type | Collagen |
| Endotoxin Level | <20.0EU per 1μg of the protein by the LAL method |
| Formulation | Type I Collagen provided at 5mg/mL (Sircol Assay). Sterility Testing: No bacterial or fungal growth detected after incubation at 37°C for 14 days following USP XXIV Chapter 71 sterility test. No mycoplasma contamination detected by PCR. |
| Quantity | 50 mg |
| Source | Fetal bovine extensor tendon-derived |
For Research Use Only. Not For Use In Diagnostic Procedures
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
