Learn More
R&D Systems™ Cultrex Collagen I (Bovine)
Description
- Collagens comprise a large family of insoluble extracellular glycoproteins that are essential components of connective tissues such as tendons, ligaments, cartilage, bone and skin.
- The mature polypeptides are secreted as coiled, left-handed helices that subsequently assemble into rope-like collagen fibers.
- Collagen I is a fibril-forming collagen that requires N- and C- terminal processing.
Specifications
Specifications
| Gene ID (Entrez) | 1277 |
| Quantity | 1 mL |
| Storage Requirements | Product is stable for a minimum of 3 months if stored at 4°C. Do Not Freeze. |
| Gene Alias | Collagen 1, Collagen I |
| Species | Bovine |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.