Learn More
R&D Systems™ Cultrex In Vitro Angiogenesis Assay (Tube Formation)
SDP
Catalog No. 3470096K
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
3470096K Each
1 options
This item is not returnable. View return policy
Catalog No. 3470096K Supplier R&D Systems™ Supplier No. 3470096K
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Includes
BME Reduced Growth Factor, 5mL; Calcein AM, 50ug; Cell Staining Solution, 15mL; 10mM Sulforaphane, 15uL
Provides a high-throughput screening platform to test the effects of multiple inhibitors and inducers on endothelial cell tube formation
Angiogenesis is the process by which new capillaries arise from pre-existing vasculature. Pathogenic angiogenesis is a common feature of rheumatoid arthritis, age-related macular degeneration, atherosclerosis and cancer.
Quantitatively assesses the ability of endothelial cells to form a tube network in a reconstituted protein matrix
Calcein AM fluorescence when cleaved to form calcein, providing data on cell viability and tube network formation in real time
Cultrex Redu
Specifications
Content And Storage The product requires storage at -20 to -70° C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Quantity Each