Learn More
Description
- Used as a substrate for cell culture adhesion
- The recommended working concentration is 1-10 μg/cm2 (1-10 μg/mL) of growth surface depending on cell type
- Supplied as 2 vials each containing 0.5 mg of protein
- Source: Murine Engelbreth-Holm-Swarm (EHS) tumor
- Storage Buffer: 50 mM Hydrochloric Acid, 1.0 mM TCEP
- Product is stable for 3 months from date of shipment when stored at ≤ -70 °C
- For optimal stability, store in liquid nitrogen; Avoid freeze-thaw cycles
- FOR LABORATORY RESEARCH USE ONLY. NOT FOR USE IN DIAGNOSTIC PROCEDURES
Specifications
Specifications
| Content And Storage | 2 vials (0.5 mg/vial). Store at -80 °C in a manual defrost freezer if using the product within 3 months. Avoid repeated freeze-thaw cycles. For optimal stability, store in liquid nitrogen. |
| Product Line | R&D Systems |
| Quantity | 1 mg |
| Source | Murine Engelbreth-Holm-Swarm (EHS) tumor |
Results may vary due to variations among tissue/cells derived from different donors or sources
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
