Learn More
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | 1 mg/mL at purity of =90% (SDS-Page). Storage Buffer: Dulbecco's Modified Eagle's Medium (DMEM) with 10 μg/mL gentamicin sulfate. |
| Gene ID (Entrez) | 284217 |
| Quantity | 1 mg |
| Storage Requirements | Product is stable for a minimum of 3 months from date of shipment when stored at -20°C in a manual defrost freezer. For optimal stability, store at -80°C. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <20.0 EU per 1μg of the protein by the LAL method. |
| Gene Alias | LAMA, LAMA1, laminin subunit alpha 1, Laminin1, Laminin-111, PTBHS, S-LAM-alpha |
| Species | Mouse |
| Product Line | R&D Systems |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

