Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Cultrex Poly-D-Lysine (2 x 100 mL)
SDP

Catalog No. 34392000101 Shop All Novus Products
Change view
Click to view available options
Quantity:
200 mL (2 x 100 mL)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-392-000101 200 mL (2 x 100 mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 34-392-000101 Supplier R&D Systems™ Supplier No. 343920001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications: Bioactivity

Substrate for cell culture adhesion. An area of 25 cm 2 can be coated with 0.5 ml of a 0.01% Poly-D-Lysine solution. Optimal conditions for attachment must be determined for each cell line and application. Slides may be dipped in the solution and air dried before applying sample.

Specifications

Content And Storage Product is stable for at least 6 months from the date of receipt when stored at 2 - 8° C. Keep sterile.
Form Liquid
Product Type Poly-D-Lysine
Sterility Sterile-filtered
Endotoxin Level <20.0 EU per 1μg of the protein by the LAL method
Formulation 0.01% in phosphate-buffered saline (PBS), sterile-filtered.
For Use With (Application) Bioactivity
Product Line R&D Systems
Quantity 200 mL (2 x 100 mL)
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.