Learn More
Description
Order Info
The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | Product is stable for at least 6 months from the date of receipt when stored at 2 - 8° C. Keep sterile. |
| Product Type | Poly-L-Lysine |
| Sterility | Sterile-filtered |
| Endotoxin Level | <20.0EU per 1μg of the protein by the LAL method |
| Formulation | 0.01% in Phosphate Buffered Saline, sterile-filtered. |
| For Use With (Application) | Bioactivity |
| Product Line | R&D Systems |
| Quantity | 100 mL |
| Regulatory Status | RUO |
| Source | Fermentation using starch. |
For research use only. Not for use in diagnostic procedures
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

