Learn More
Description
- Poly-L-Ornithine along or in combination with Fibronectin or Laminin-I is frequently used to enhance attachment and differentiation of various types of neuronal and neural stem cells.
Specifications
Specifications
| Quantity | 100 mL |
| Storage Requirements | Product is stable for at least 6 months from the date of receipt when stored at 2 - 8°C. Keep sterile. |
| Species | Multi-species |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
