Learn More
Specifications
Specifications
| Content And Storage | Product is stable for a minimum of 3 months if stored at 4° C. Do Not Freeze. |
| Concentration | 3 mg/mL |
| Endotoxin Level | <20EU/mL |
| Formulation | Type I Collagen is provided at 3mg/mL (Sircol Assay). |
| For Use With (Application) | Bioactivity |
| Product Line | R&D Systems |
| Quantity | 35 mL |
| Regulatory Status | RUO |
| Source | Rat tail tendons. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
