Learn More
R&D Systems™ Cultrex RGF Basement Membrane Extract, Type R1
Description
- Qualified for use in organoid cell culture.
- Improves take-rate and growth in difficult organoid systems.
- Rich in Entactin.
- Quality controlled for peformance consistency.
Specifications
Specifications
| Concentration | 8-12 mg/mL (by Lowry assay) |
| Content And Storage | Product is stable for a minimum of 3 months from date of shipment when stored at -20°C in a manual defrost freezer. For optimal stability, store (at -80°C. Avoid freeze-thaw cycles.), Store the unopened product at -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. |
| Quantity | 1 mL |
| Source | Murine Engelbreth-Holm-Swarm (EHS) tumor |
| Species | Mouse, Multi-Species |
| Description | For optimal stability, store at -80°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.